Assay Details
| Gene Symbol: Tango2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC005445.1 | - | 1 - 2 | 100 | 3496749 | 67 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC005445.1 transcripts.
Set Membership: |
|

Tango2
Genomic Map