Assay Details
| Gene Symbol: Klhl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086748.1 | - | 1 - 2 | 32 | 52 | |
| CJ159851.1 | - | 1 - 2 | 77 | 52 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl22| Gene Symbol: Klhl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086748.1 | - | 1 - 2 | 32 | 52 | |
| CJ159851.1 | - | 1 - 2 | 77 | 52 | ||
Set Membership: |
|