Assay Details
| Gene Symbol: Lrrc74b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029053.3 | NP_083329.2 | 8 - 9 | 1047 | 72 | |
| GenBank mRNA | AK015438.1 | - | 5 - 6 | 1521 | 72 | |
| BC115678.1 | - | 8 - 9 | 1024 | 40104166 | 72 | |
| BC115679.1 | - | 8 - 9 | 1024 | 40104167 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc74b
Genomic Map