Assay Details
| Gene Symbol: Uso1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019490.1 | NP_062363.1 | 19 - 20 | 2450 | 85 | |
| GenBank mRNA | AF096868.1 | - | 18 - 19 | 2238 | 85 | |
| AK168433.1 | - | 7 - 8 | 981 | 85 | ||
| BC005548.1 | - | 3 - 4 | 418 | 85 | ||
| BC016069.1 | - | 19 - 20 | 2450 | 4480992 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uso1
Genomic Map