Assay Details
| Gene Symbol: Cxcl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008599.4 | NP_032625.2 | 1 - 2 | 119 | 63 | |
| GenBank mRNA | AK042017.1 | - | 1 - 2 | 121 | 63 | |
| AK042480.1 | - | 1 - 2 | 37 | 63 | ||
| AK156012.1 | - | 1 - 2 | 120 | 63 | ||
| AK156029.1 | - | 1 - 2 | 120 | 63 | ||
| AK156453.1 | - | 1 - 2 | 120 | 63 | ||
| AK157109.1 | - | 1 - 2 | 120 | 63 | ||
| AK157118.1 | - | 1 - 2 | 120 | 63 | ||
| BC003343.1 | - | 1 - 2 | 107 | 3257716 | 63 | |
| CT010194.1 | - | 1 - 2 | 63 | 63 | ||
| M34815.1 | - | 1 - 2 | 157 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl9
Genomic Map