| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500024211.mm.1 |
TC0500002745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024211.mm.1 |
NM_001080794.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024211.mm.1 |
NM_001080795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024211.mm.1 |
NM_001080796.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024211.mm.1 |
NM_011816.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024208.mm.1 |
TC0500002745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024208.mm.1 |
NM_001080794.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024208.mm.1 |
NM_001080795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024208.mm.1 |
NM_001080796.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024208.mm.1 |
NM_001080797.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024208.mm.1 |
NM_011816.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024213.mm.1 |
TC0500002745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024213.mm.1 |
NM_001080795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024213.mm.1 |
NM_011816.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024212.mm.1 |
TC0500002745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024212.mm.1 |
NM_001080795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024212.mm.1 |
NM_011816.4 |
Refseq |