Assay Details
| Gene Symbol: Zcchc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138590.4 | NP_613056.2 | 6 - 7 | 1133 | 97 | |
| GenBank mRNA | BC050200.1 | - | 5 - 6 | 1008 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zcchc7
Genomic Map