Assay Details
| Gene Symbol: Pax5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008782.2 | NP_032808.1 | 6 - 7 | 808 | 81 | |
| GenBank mRNA | JX065130.1 | - | 5 - 6 | 689 | 81 | |
| JX065131.1 | - | 1 - 2 | 146 | 81 | ||
| M97013.1 | - | 6 - 7 | 808 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pax5
Genomic Map