Assay Details
| Gene Symbol: Erap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030711.4 | NP_109636.1 | 13 - 14 | 1986 | 82 | |
| GenBank mRNA | AB047552.1 | - | 12 - 13 | 1974 | 82 | |
| AF227511.1 | - | 12 - 13 | 1962 | 82 | ||
| AK029841.1 | - | 13 - 14 | 1985 | 82 | ||
| AK030329.1 | - | 13 - 14 | 1987 | 82 | ||
| AK036559.1 | - | 1987 | 82 | |||
| AK040350.1 | - | 10 - 11 | 1204 | 82 | ||
| AK159406.1 | - | 13 - 14 | 1987 | 82 | ||
| BC046610.1 | - | 13 - 14 | 1992 | 6397585 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erap1
Genomic Map