Assay Details
| Gene Symbol: Slc6a20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139142.2 | NP_631881.1 | 2 - 3 | 292 | 63 | |
| GenBank mRNA | AJ428067.1 | - | 305 | 63 | ||
| AJ964961.1 | - | 2 - 3 | 266 | 63 | ||
| AK043297.1 | - | 2 - 3 | 303 | 63 | ||
| AK080979.1 | - | 303 | 63 | |||
| AK140445.1 | - | 303 | 63 | |||
| AK160468.1 | - | 303 | 63 | |||
| BC118933.2 | - | 2 - 3 | 292 | 40090800 | 63 | |
More Information
Set Membership: |
|

Slc6a20a
Genomic Map