Assay Details
| Gene Symbol: Ncbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033201.3 | NP_001028373.2 | 12 - 13 | 1573 | 91 | |
| GenBank mRNA | AK133991.1 | - | 12 - 13 | 1529 | 91 | |
| AK134334.1 | - | 12 - 13 | 1564 | 91 | ||
| BC033473.1 | - | 11 - 12 | 1189 | 91 | ||
| BC034658.1 | - | 9 - 10 | 1015 | 91 | ||
| BC055777.1 | - | 9 - 10 | 999 | 91 | ||
| BC138898.1 | - | 12 - 13 | 1444 | 8861920 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncbp1
Genomic Map