| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400003638.mm.1 |
NM_001033201.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003638.mm.1 |
NM_001033201.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003638.mm.1 |
TC0400000501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400003637.mm.1 |
NM_001033201.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003637.mm.1 |
NM_001033201.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003637.mm.1 |
TC0400000501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400003636.mm.1 |
NM_001033201.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003636.mm.1 |
NM_001033201.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003636.mm.1 |
TC0400000501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400003639.mm.1 |
NM_001033201.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003639.mm.1 |
NM_001033201.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400003639.mm.1 |
TC0400000501.mm.1 |
MTA TC |