Assay Details
| Gene Symbol: Ncbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033201.3 | NP_001028373.2 | 21 - 22 | 2484 | 80 | |
| GenBank mRNA | AK133991.1 | - | 21 - 22 | 2440 | 80 | |
| AK134334.1 | - | 21 - 22 | 2475 | 80 | ||
| BC033473.1 | - | 20 - 21 | 2100 | 80 | ||
| BC034658.1 | - | 18 - 19 | 1926 | 80 | ||
| BC055777.1 | - | 18 - 19 | 1910 | 80 | ||
| BC138898.1 | - | 21 - 22 | 2355 | 8861920 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncbp1
Genomic Map