Assay Details
| Gene Symbol: Ncbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033201.3 | NP_001028373.2 | 2 - 3 | 465 | 101 | |
| GenBank mRNA | AK133991.1 | - | 2 - 3 | 421 | 101 | |
| AK134334.1 | - | 2 - 3 | 456 | 101 | ||
| AK166458.1 | - | 2 - 3 | 247 | 101 | ||
| AK169557.1 | - | 2 - 3 | 466 | 101 | ||
| BC033473.1 | - | 1 - 2 | 81 | 101 | ||
| BC138898.1 | - | 2 - 3 | 336 | 8861920 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncbp1
Genomic Map