Assay Details
| Gene Symbol: Vmn1r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039747.1 | - | 2 - 3 | 1551 | 86 | |
| AK082576.1 | - | 3 - 4 | 497 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r4| Gene Symbol: Vmn1r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039747.1 | - | 2 - 3 | 1551 | 86 | |
| AK082576.1 | - | 3 - 4 | 497 | 86 | ||
Set Membership: |
|