Assay Details
| Gene Symbol: Tmeff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ400622.1 | - | 4 - 5 | 381 | 94 | |
| AK043792.1 | - | 2 - 3 | 88 | 94 | ||
| AK079633.1 | - | 2 - 3 | 68 | 94 | ||
More Information
Set Membership: |
|

Tmeff1
Genomic Map