Assay Details
| Gene Symbol: Tmeff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021436.2 | NP_067411.1 | 8 - 9 | 1146 | 80 | |
| GenBank mRNA | AJ400622.1 | - | 8 - 9 | 815 | 80 | |
| AK043792.1 | - | 6 - 7 | 522 | 80 | ||
| AK079633.1 | - | 6 - 7 | 496 | 80 | ||
| BC023714.1 | - | 3 - 4 | 308 | 80 | ||
| BC057598.1 | - | 8 - 9 | 1146 | 6401547 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmeff1
Genomic Map