Assay Details
| Gene Symbol: Akap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB028920.1 | - | 4 - 5 | 411 | 139 | |
| AK004801.1 | - | 4 - 5 | 417 | 139 | ||
| AK089092.1 | - | 4 - 5 | 424 | 139 | ||
| BC042602.1 | - | 4 - 5 | 407 | 139 | ||
| BC060170.1 | - | 4 - 5 | 402 | 139 | ||
| BC078448.1 | - | 4 - 5 | 395 | 139 | ||
| BC125521.1 | - | 4 - 5 | 407 | 40130036 | 139 | |
| BC137746.1 | - | 4 - 5 | 407 | 8860765 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap8
Genomic Map