Assay Details
| Gene Symbol: Akap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB028920.1 | - | 5 - 6 | 891 | 60 | |
| AK004801.1 | - | 5 - 6 | 897 | 60 | ||
| AK089092.1 | - | 5 - 6 | 904 | 60 | ||
| BC042602.1 | - | 5 - 6 | 885 | 60 | ||
| BC054433.1 | - | 3 - 4 | 583 | 60 | ||
| BC060170.1 | - | 5 - 6 | 882 | 60 | ||
| BC060209.1 | - | 4 - 5 | 1387 | 60 | ||
| BC125521.1 | - | 5 - 6 | 887 | 40130036 | 60 | |
| BC137746.1 | - | 5 - 6 | 887 | 8860765 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap8
Genomic Map