Assay Details
| Gene Symbol: Akap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004801.1 | - | 6 - 7 | 1033 | 101 | |
| AK089092.1 | - | 6 - 7 | 1040 | 101 | ||
| BC042602.1 | - | 6 - 7 | 1021 | 101 | ||
| BC054433.1 | - | 4 - 5 | 719 | 101 | ||
| BC060170.1 | - | 6 - 7 | 1018 | 101 | ||
| BC060209.1 | - | 5 - 6 | 1523 | 101 | ||
| BC078448.1 | - | 5 - 6 | 1413 | 101 | ||
| BC125521.1 | - | 6 - 7 | 1023 | 40130036 | 101 | |
| BC137746.1 | - | 6 - 7 | 1023 | 8860765 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap8
Genomic Map