Assay Details
| Gene Symbol: Akap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB028920.1 | - | 7 - 8 | 1065 | 66 | |
| AK004801.1 | - | 7 - 8 | 1071 | 66 | ||
| AK089092.1 | - | 7 - 8 | 1078 | 66 | ||
| BC042602.1 | - | 7 - 8 | 1059 | 66 | ||
| BC054433.1 | - | 5 - 6 | 757 | 66 | ||
| BC060170.1 | - | 7 - 8 | 1056 | 66 | ||
| BC060209.1 | - | 6 - 7 | 1561 | 66 | ||
| BC078448.1 | - | 6 - 7 | 1451 | 66 | ||
| BC125521.1 | - | 7 - 8 | 1061 | 40130036 | 66 | |
| BC137746.1 | - | 7 - 8 | 1061 | 8860765 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap8
Genomic Map