Assay Details
| Gene Symbol: Akap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB028920.1 | - | 8 - 9 | 1097 | 77 | |
| AK004801.1 | - | 8 - 9 | 1103 | 77 | ||
| AK089092.1 | - | 8 - 9 | 1110 | 77 | ||
| BC060209.1 | - | 7 - 8 | 1593 | 77 | ||
| BC078448.1 | - | 7 - 8 | 1483 | 77 | ||
| BC125521.1 | - | 8 - 9 | 1093 | 40130036 | 77 | |
| BC137746.1 | - | 8 - 9 | 1093 | 8860765 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap8
Genomic Map