Assay Details
| Gene Symbol: Rrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163734.1 | NP_001157206.1 | 8 - 9 | 980 | 75 | |
| NM_028244.2 | NP_082520.2 | 11 - 12 | 1296 | 75 | ||
| GenBank mRNA | AK011155.1 | - | 4 - 5 | 222 | 75 | |
| AK129078.1 | - | 8 - 9 | 980 | 75 | ||
| BC016569.1 | - | 11 - 12 | 1296 | 3157173 | 75 | |
More Information
Set Membership: |
|

Rrp1b
Genomic Map