Assay Details
| Gene Symbol: Rrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163734.1 | NP_001157206.1 | 2 - 3 | 420 | 102 | |
| NM_028244.2 | NP_082520.2 | 5 - 6 | 736 | 102 | ||
| GenBank mRNA | AK129078.1 | - | 2 - 3 | 420 | 102 | |
| BC016569.1 | - | 5 - 6 | 736 | 3157173 | 102 | |
More Information
Set Membership: |
|

Rrp1b
Genomic Map