Assay Details
| Gene Symbol: Ephx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033163.3 | NP_001028335.1 | 2 - 3 | 537 | 76 | |
| GenBank mRNA | AK010021.1 | - | 2 - 3 | 524 | 76 | |
| AK132483.1 | - | 2 - 3 | 538 | 76 | ||
| BC145797.1 | - | 2 - 3 | 602 | 40131100 | 76 | |
More Information
Set Membership: |
|

Ephx3
Genomic Map