Assay Details
| Gene Symbol: Sik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010831.2 | NP_034961.2 | 4 - 5 | 471 | 121 | |
| GenBank mRNA | AK141817.1 | - | 4 - 5 | 471 | 121 | |
| AK172427.1 | - | 4 - 5 | 471 | 121 | ||
| U11494.2 | - | 4 - 5 | 463 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sik1
Genomic Map