Assay Details
| Gene Symbol: Akap8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017476.2 | NP_059504.2 | 10 - 11 | 1381 | 74 | |
| GenBank mRNA | AB028921.1 | - | 10 - 11 | 1377 | 74 | |
| AK011325.1 | - | 10 - 11 | 1386 | 74 | ||
| AK166526.1 | - | 11 - 12 | 1297 | 74 | ||
| BC046801.1 | - | 10 - 11 | 1370 | 6403966 | 74 | |
| BC051015.1 | - | 8 - 9 | 3690 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akap8l
Genomic Map