Assay Details
| Gene Symbol: Akap8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017476.2 | NP_059504.2 | 11 - 12 | 1484 | 98 | |
| GenBank mRNA | AB028921.1 | - | 11 - 12 | 1480 | 98 | |
| AK011325.1 | - | 11 - 12 | 1489 | 98 | ||
| AK166526.1 | - | 12 - 13 | 1400 | 98 | ||
| BC003242.1 | - | 1 - 2 | 113 | 98 | ||
| BC046801.1 | - | 11 - 12 | 1473 | 6403966 | 98 | |
| BC051015.1 | - | 9 - 10 | 3793 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akap8l
Genomic Map