Assay Details
| Gene Symbol: Akap8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017476.2 | NP_059504.2 | 1 - 2 | 88 | 64 | |
| GenBank mRNA | AB028921.1 | - | 1 - 2 | 81 | 64 | |
| AK011325.1 | - | 1 - 2 | 93 | 64 | ||
| AK166526.1 | - | 1 - 2 | 106 | 64 | ||
| BC010401.1 | - | 1 - 2 | 138 | 64 | ||
| BC046801.1 | - | 1 - 2 | 77 | 6403966 | 64 | |
| BC051015.1 | - | 1 - 2 | 88 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB028921.1, AK011325.1, AK166526.1, BC046801.1, NM_017476.2 transcripts.
Set Membership: |
|

Akap8l
Genomic Map