Assay Details
| Gene Symbol: Wdr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027113.2 | NP_081389.1 | 1 | 1727 | 130 | |
| GenBank mRNA | AK009247.1 | - | 1 | 1642 | 130 | |
| AK149554.1 | - | 1698 | 130 | |||
| AK165100.1 | - | 1591 | 130 | |||
| BC064045.1 | - | 1 | 1727 | 1245342 | 130 | |
| EF011616.1 | - | 1 | 1725 | 130 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673431 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | BC064045.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | EF011616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | AK149554.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | AK165100.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | AK009247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | BC064045.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | EF011616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | AK149554.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | AK165100.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P281711 | AK009247.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937903 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141628 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129596 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173204 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | BC064045.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | EF011616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | AK149554.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | AK165100.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | AK009247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | BC064045.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | EF011616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | AK149554.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | AK165100.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P281711 | AK009247.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30673407 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173204 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129596 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141628 | - | - |
| Whole Mouse Genome G4122A | A_52_P281711 | BC064045.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P281711 | EF011616.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P281711 | AK149554.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P281711 | AK165100.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P281711 | AK009247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P281711 | BC064045.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P281711 | EF011616.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P281711 | AK149554.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P281711 | AK165100.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P281711 | AK009247.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103506 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141628 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173204 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129596 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12624073 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12836308 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12836309 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12624079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12624053 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30673428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30673397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12624061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12624083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12872511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12895430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12872510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12624079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12685388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30673392 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10435488 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453058_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10435487 | - | - |
| Mouse Genome 430 2.0 Array | 1453058_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453058_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69544_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69544_at | - | - |
| Mouse Expression Set 430 | 1453058_PM_at | - | - |
| Mouse Expression Set 430 | 1453058_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453058_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453058_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 571305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 670110 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 670110 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000670110 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2425488 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2425488 | - | - |
| mouse-6 v1.1 (Illumina) | scl49184.1.1_144-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 670110 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49184.1.1_144-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670110 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2425488 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2425488 | - | - |

Wdr5b
Genomic Map