Assay Details
| Gene Symbol: Smim22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253796.1 | NP_001240725.1 | 2 - 3 | 245 | 147 | |
| NM_001253797.1 | NP_001240726.1 | 2 - 3 | 245 | 132 | ||
| NM_001253802.1 | NP_001240731.1 | 2 - 3 | 180 | 147 | ||
| NM_001253803.1 | NP_001240732.1 | 2 - 3 | 180 | 132 | ||
| NR_045595.1 | 2 - 3 | 215 | 147 | |||
| GenBank mRNA | AW985536.1 | - | 2 - 3 | 171 | 132 | |
| BB610432.1 | - | 1 - 2 | 82 | 132 | ||
| BC047216.1 | - | 2 - 3 | 224 | 147 | ||
| CJ043101.1 | - | 2 - 3 | 181 | 147 | ||
| CJ237462.1 | - | 2 - 3 | 242 | 132 | ||
| CJ320043.1 | - | 2 - 3 | 242 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smim22
Genomic Map