Assay Details
| Gene Symbol: Zkscan16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099323.2 | NP_001092793.1 | 3 - 4 | 658 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zkscan16| Gene Symbol: Zkscan16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099323.2 | NP_001092793.1 | 3 - 4 | 658 | 82 | |
Set Membership: |
|