Assay Details
| Gene Symbol: Zkscan16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099323.2 | NP_001092793.1 | 5 - 6 | 894 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059502 | NM_001099323.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371213 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481412 | NM_146394.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57393 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059502 | NM_001099323.2 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22953793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03451630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03284468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03214284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04046708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03452957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03735479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03385464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03961977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03843753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04063840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04085997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04063844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03690611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03975074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03665822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04046709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04046701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03255833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22953713 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10505169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505164 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505166 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435407_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10505163 | - | - |
| Mouse Genome 430 2.0 Array | 1435407_at | - | - |
| Mouse Genome 430 2.0 Array | 1435407_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400004597.mm.1 | NM_001099323.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400004597.mm.1 | NM_001099323.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400004597.mm.1 | TC0400000650.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435407_PM_at | - | - |
| Mouse Expression Set 430 | 1435407_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435407_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435407_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 399510 | - | - |

Zkscan16
Genomic Map