Assay Details
| Gene Symbol: Rbm24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081425.1 | NP_001074894.1 | 3 - 4 | 1078 | 57 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CJ159111.1 | - | 246 | Chr. - - [-] on Build | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P920158 | NM_001081425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P920158 | NM_001081425.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573845 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708823 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3005 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P920158 | NM_001081425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P920158 | NM_001081425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P920158 | NM_001081425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P920158 | NM_001081425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487832 | NM_001081425.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29007810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10537019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10930334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10521726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10537012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10605871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10559695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10915665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10671306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10537031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10591708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10525979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10599959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10847788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10599952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10682609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10682603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10599958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10512646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10729879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10599957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11005632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11017913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10688776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10537025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29007645 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404909 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404908 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404906 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404905 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404907 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456491_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443715_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456180_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458624_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454752_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404904 | - | - |
| Mouse Genome 430 2.0 Array | 1443715_at | - | - |
| Mouse Genome 430 2.0 Array | 1454752_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458624_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454752_at | - | - |
| Mouse Genome 430 2.0 Array | 1443715_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458624_at | - | - |
| Mouse Genome 430 2.0 Array | 1456491_at | - | - |
| Mouse Genome 430 2.0 Array | 1456491_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 666794_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 666794_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300003251.mm.1 | NM_001081425.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003251.mm.1 | NM_001081425.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003251.mm.1 | TC1300000532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300003248.mm.1 | NM_001081425.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003248.mm.1 | NM_001081425.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003248.mm.1 | TC1300000532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300003249.mm.1 | NM_001081425.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003249.mm.1 | NM_001081425.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003249.mm.1 | TC1300000532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300003250.mm.1 | NM_001081425.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003250.mm.1 | NM_001081425.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003250.mm.1 | TC1300000532.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1443715_at | - | - |
| Mouse Expression Set 430 | 1454752_at | - | - |
| Mouse Expression Set 430 | 1454752_PM_at | - | - |
| Mouse Expression Set 430 | 1456491_PM_at | - | - |
| Mouse Expression Set 430 | 1456491_at | - | - |
| Mouse Expression Set 430 | 1458624_PM_at | - | - |
| Mouse Expression Set 430 | 1443715_PM_at | - | - |
| Mouse Expression Set 430 | 1458624_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443715_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454752_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456491_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456491_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443715_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458624_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458624_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456180_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456180_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454752_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 319380 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246156 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2802711 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2802711 | - | - |

Rbm24
Genomic Map