Assay Details
| Gene Symbol: Slc35a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083937.1 | NP_001077406.1 | 2 - 3 | 369 | 61 | |
| NM_078484.2 | NP_511039.2 | 2 - 3 | 369 | 61 | ||
| GenBank mRNA | AB027147.1 | - | 2 - 3 | 288 | 61 | |
| AF229634.1 | - | 2 - 3 | 369 | 61 | ||
| AK008095.1 | - | 1 - 2 | 92 | 61 | ||
| AK146830.1 | - | 2 - 3 | 285 | 61 | ||
| AK155755.1 | - | 2 - 3 | 281 | 61 | ||
| BC037701.1 | - | 2 - 3 | 306 | 4187325 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35a2
Genomic Map