Assay Details
| Gene Symbol: Diap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172493.2 | NP_766081.1 | 12 - 13 | 1573 | 105 | |
| GenBank mRNA | AB713187.1 | - | 13 - 14 | 1368 | 105 | |
| AK083254.1 | - | 5 - 6 | 505 | 105 | ||
| AK088648.1 | - | 12 - 13 | 1507 | 105 | ||
| AK153986.1 | - | 12 - 13 | 1561 | 105 | ||
| AY312280.1 | - | 12 - 13 | 1510 | 105 | ||
More Information
Set Membership: |
|

Diap2
Genomic Map