Assay Details
| Gene Symbol: Diap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172493.2 | NP_766081.1 | 13 - 14 | 1691 | 127 | |
| GenBank mRNA | AB713187.1 | - | 14 - 15 | 1486 | 127 | |
| AK083254.1 | - | 6 - 7 | 623 | 127 | ||
| AK088648.1 | - | 13 - 14 | 1625 | 127 | ||
| AK153986.1 | - | 13 - 14 | 1679 | 127 | ||
| AY312280.1 | - | 13 - 14 | 1628 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap2
Genomic Map