Assay Details
| Gene Symbol: Diap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172493.2 | NP_766081.1 | 22 - 23 | 2960 | 69 | |
| GenBank mRNA | AB713187.1 | - | 23 - 24 | 2755 | 69 | |
| AK083254.1 | - | 15 - 16 | 1892 | 69 | ||
| AK088648.1 | - | 22 - 23 | 2894 | 69 | ||
| AK153986.1 | - | 22 - 23 | 2948 | 69 | ||
| AY312280.1 | - | 22 - 23 | 2897 | 69 | ||
| Y15910.1 | - | 3 - 4 | 467 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap2
Genomic Map