Assay Details
| Gene Symbol: Diap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172493.2 | NP_766081.1 | 7 - 8 | 980 | 141 | |
| GenBank mRNA | AB713187.1 | - | 8 - 9 | 775 | 141 | |
| AK088648.1 | - | 7 - 8 | 914 | 141 | ||
| AK153986.1 | - | 7 - 8 | 968 | 141 | ||
| AY312280.1 | - | 7 - 8 | 917 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap2
Genomic Map