Assay Details
| Gene Symbol: Rassf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028478.2 | NP_082754.2 | 3 - 4 | 285 | 129 | |
| GenBank mRNA | AK005472.1 | - | 3 - 4 | 272 | 129 | |
| AK132288.1 | - | 3 - 4 | 286 | 129 | ||
| BC031808.1 | - | 3 - 4 | 240 | 129 | ||
| BC039934.1 | - | 3 - 4 | 296 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rassf6
Genomic Map