Assay Details
| Gene Symbol: Rassf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028478.2 | NP_082754.2 | 8 - 9 | 866 | 70 | |
| GenBank mRNA | AK005472.1 | - | 8 - 9 | 814 | 70 | |
| AK132288.1 | - | 8 - 9 | 828 | 70 | ||
| BC031808.1 | - | 8 - 9 | 838 | 70 | ||
| BC039934.1 | - | 8 - 9 | 913 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rassf6
Genomic Map