Assay Details
| Gene Symbol: Rassf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028478.2 | NP_082754.2 | 9 - 10 | 988 | 95 | |
| GenBank mRNA | AK005472.1 | - | 9 - 10 | 936 | 95 | |
| AK132288.1 | - | 9 - 10 | 950 | 95 | ||
| BC031808.1 | - | 9 - 10 | 960 | 95 | ||
| BC039934.1 | - | 9 - 10 | 1035 | 95 | ||
| BC075725.1 | - | 6 - 7 | 2326 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rassf6
Genomic Map