Assay Details
| Gene Symbol: Slc4a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136260.1 | NP_001129732.1 | 5 - 6 | 621 | 71 | |
| NM_001197147.1 | NP_001184076.1 | 5 - 6 | 621 | 71 | ||
| NM_018760.2 | NP_061230.2 | 5 - 6 | 621 | 71 | ||
| GenBank mRNA | AF141934.3 | - | 2 - 3 | 423 | 71 | |
| AK140443.1 | - | 5 - 6 | 1064 | 71 | ||
| HQ018820.1 | - | 2 - 3 | 458 | 71 | ||
| HQ204220.1 | - | 5 - 6 | 592 | 71 | ||
| HQ285250.1 | - | 2 - 3 | 461 | 71 | ||
More Information
Set Membership: |
|

Slc4a4
Genomic Map