Assay Details
| Gene Symbol: Slc4a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136260.1 | NP_001129732.1 | 9 - 10 | 1114 | 73 | |
| NM_001197147.1 | NP_001184076.1 | 9 - 10 | 1087 | 73 | ||
| NM_018760.2 | NP_061230.2 | 9 - 10 | 1114 | 73 | ||
| GenBank mRNA | AF141934.3 | - | 6 - 7 | 916 | 73 | |
| AK140443.1 | - | 9 - 10 | 1557 | 73 | ||
| BC129863.1 | - | 9 - 10 | 1076 | 73 | ||
| BC129864.1 | - | 9 - 10 | 1075 | 73 | ||
| BC148292.2 | - | 9 - 10 | 1050 | 40110074 | 73 | |
| HQ018820.1 | - | 6 - 7 | 951 | 73 | ||
| HQ204220.1 | - | 9 - 10 | 1058 | 73 | ||
| HQ285250.1 | - | 6 - 7 | 927 | 73 | ||
More Information
Set Membership: |
|

Slc4a4
Genomic Map