Assay Details
| Gene Symbol: Rp2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290643.1 | NP_001277572.1 | 5 - 6 | 1306 | 120 | |
| GenBank mRNA | AK132713.1 | - | 5 - 6 | 1306 | 120 | |
| BC049698.1 | - | 5 - 6 | 1184 | 6706696 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rp2h
Genomic Map