Assay Details
| Gene Symbol: Rp2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110967.1 | 6 - 7 | 1382 | 127 | ||
| GenBank mRNA | AK076907.1 | - | 6 - 7 | 1244 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Rp2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110967.1 | 6 - 7 | 1382 | 127 | ||
| GenBank mRNA | AK076907.1 | - | 6 - 7 | 1244 | 127 | |
Set Membership: |
|