Assay Details
| Gene Symbol: Ftsj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290430.1 | NP_001277359.1 | 3 - 4 | 465 | 90 | |
| NM_133991.2 | NP_598752.2 | 3 - 4 | 465 | 90 | ||
| GenBank mRNA | AK004357.1 | - | 3 - 4 | 423 | 90 | |
| AK036313.1 | - | 3 - 4 | 423 | 90 | ||
| BC034880.1 | - | 3 - 4 | 335 | 3468633 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ftsj1
Genomic Map