Assay Details
| Gene Symbol: Ftsj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290430.1 | NP_001277359.1 | 4 - 5 | 554 | 76 | |
| NM_133991.2 | NP_598752.2 | 4 - 5 | 554 | 76 | ||
| GenBank mRNA | AK004357.1 | - | 4 - 5 | 512 | 76 | |
| AK036313.1 | - | 4 - 5 | 512 | 76 | ||
| AK150251.1 | - | 3 - 4 | 438 | 76 | ||
| BC034880.1 | - | 4 - 5 | 424 | 3468633 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ftsj1
Genomic Map