Assay Details
| Gene Symbol: Porcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB036746.1 | - | 3 - 4 | 506 | 74 | |
| AB036747.1 | - | 3 - 4 | 506 | 74 | ||
| AB036748.1 | - | 3 - 4 | 506 | 74 | ||
| AB036749.1 | - | 3 - 4 | 506 | 74 | ||
| BC027292.1 | - | 3 - 4 | 387 | 74 | ||
| BC032284.1 | - | 3 - 4 | 476 | 5362485 | 74 | |
| BF462782.1 | - | 3 - 4 | 499 | 74 | ||
| BQ556448.1 | - | 2 - 3 | 323 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Porcn
Genomic Map