Assay Details
Gene Symbol: Zfp799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293229.1 | NP_001280158.1 | 1 - 2 | 172 | 115 | |
NM_177359.5 | NP_796333.3 | 1 - 2 | 172 | 115 | ||
GenBank mRNA | AK031686.1 | - | 1 - 2 | 160 | 115 | |
AK083640.1 | - | 1 - 2 | 149 | 115 | ||
AK137037.1 | - | 1 - 2 | 149 | 115 | ||
BC062813.1 | - | 1 - 2 | 126 | 115 | ||
BC096552.1 | - | 1 - 2 | 122 | 30618827 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK031686.1, AK083640.1, AK137037.1, BC062813.1, BC096552.1, NM_001293229.1, NM_177359.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
